Anti-RBL2 Rabbit Monoclonal Antibody (Unconjugated) [clone: 12.A1.1]

Lieferant: Rockland Immunochemicals
ROCK100-401-173EA 783 EUR
ROCK100-401-173
Anti-RBL2 Rabbit Monoclonal Antibody (Unconjugated) [clone: 12.A1.1]
Antikörper
Suitable for ELISA, immunoprecipitation, immunoblotting, immunochemistry, and other immunological methods requiring high titer and specificity.

Recommended Dilutions: ELISA: 1:5000 - 1:20000; Western Blot: 1:500 - 1:2000; Immunochemistry: 1:200 - 1:1000; Immunoprecipitation: 1:100;

Type: Primary
Antigen: RBL2
Clonality: Monoclonal
Clone: 12.A1.1
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype:
Reactivity: Mouse, Rat, Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR