Anti-TYMS Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
ROCK100-401-199EA 379 EUR
ROCK100-401-199
Anti-TYMS Rabbit Polyclonal Antibody
Antikörper
This antiserum against Human Thymidylate Synthase (hTS) has been tested for use in ELISA, immunoprecipitation, immunofluorescence microscopy, immunochemistry and immunoblotting.

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:500 - 1:2000;

Type: Primary
Antigen: TYMS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR