Anti-MAPK1 Rabbit Monoclonal Antibody (Unconjugated) [clone: 3G2]

Lieferant: Rockland Immunochemicals
ROCK100-401-218EA 379 EUR
ROCK100-401-218
Anti-MAPK1 Rabbit Monoclonal Antibody (Unconjugated) [clone: 3G2]
Antikörper
This antiserum was tested for use in ELISA and by western blot.  Specific conditions for reactivity should be optimized by the end user. Expect a predominant band approximately 42 kDa in size corresponding to p42 MAP Kinase (ERK2) by western blotting.

Recommended Dilutions: ELISA: 1:2000 to 1:10000; Western Blot: 1:1000-1:,2000; Immunoprecipitation: 5.0 µg/mg sample;

Type: Primary
Antigen: MAPK1
Clonality: Monoclonal
Clone: 3G2
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR