Anti-CHUK Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
ROCK100-401-219EA 783 EUR
ROCK100-401-219
Anti-CHUK Rabbit Polyclonal Antibody
Antikörper
Anti-IKKa antibody was assayed by immunoblot and found to be reactive against IKK a at a dilution of 1:1000 followed by reaction with Peroxidase conjugated Affinity Purified anti-Rabbit IgG (H&L) (Goat).

Recommended Dilutions: ELISA: 1:5000 - 1:25000; Western Blot: 1:500 - 1:3000;

Type: Primary
Antigen: CHUK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Mouse, Rat, Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR