Anti-IKBKB Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
ROCK100-401-220EA 783 EUR
ROCK100-401-220
Anti-IKBKB Rabbit Polyclonal Antibody
Antikörper
This product was assayed by immunoblot and found to be reactive against IKKb at a dilution of 1:1000 followed by reaction with Peroxidase conjugated Affinity Purified anti-Rabbit IgG (H&L) (Goat).

Recommended Dilutions: ELISA: 1:1000 - 1:5000; Western Blot: 1:200 - 1:1000; Immunochemistry: 1:200 - 1:1000;

Type: Primary
Antigen: IKBKB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Mouse, Rat, Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR