Anti-GLI1 Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
ROCK100-401-223EA 379 EUR
ROCK100-401-223
Anti-GLI1 Rabbit Polyclonal Antibody
Antikörper
This antibody has been tested for use in ELISA, immunochemistry and western blot.  Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 120 kDa in size corresponding to Gli-1 Protein by western blotting.

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:500 - 1:2000;

Type: Primary
Antigen: GLI1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse, Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR