Anti-MYC Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
ROCK100-401-224EA 379 EUR
ROCK100-401-224
Anti-MYC Rabbit Polyclonal Antibody
Antikörper
This antibody reacts with human and mouse c-myc Protein, which is a nuclear Protein with a molecular weight of approximately 66 kDa.  This antiserum detects c-myc by immunoblotting assays in both overexpressed and endogenous systems.

Recommended Dilutions: ELISA: 1:10000 - 1:50000; Western Blot: 1:500- 1:2000;

Type: Primary
Antigen: MYC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR