Anti-DAB1 Rabbit Monoclonal Antibody (Unconjugated) [clone: AC240]

Lieferant: Rockland Immunochemicals
ROCK100-401-225EA 379 EUR
ROCK100-401-225
Anti-DAB1 Rabbit Monoclonal Antibody (Unconjugated) [clone: AC240]
Antikörper
Anti-Dab1 antibody is suitable for the detection of Dab1 by immunoprecipitation.

Recommended Dilutions: ELISA: 1:10000 - 1:50000; Western Blot: 1:5000; Immunoprecipitation: 1 µl per 500 µg;

Type: Primary
Antigen: DAB1
Clonality: Monoclonal
Clone: AC240
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR