Anti-MED23 Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
ROCK100-401-239EA 398 EUR
ROCK100-401-239
Anti-MED23 Rabbit Polyclonal Antibody
Antikörper
Anti-DRIP-130 detects a 130 kDa band by immunoblot analysis using a dilution of 1:1,000. For immunoprecipitation a dilution of 1:100 or 1:200 is suggested; pre-clearing with a non-specific rabbit IgG is helpful to reduce background.

Recommended Dilutions: ELISA: 1:5000 - 1:25000; Western Blot: 1:1000; Immunoprecipitation: 1:100 - 1:200;
Restore with deionized water (or equivalent); 500 µl

Type: Primary
Antigen: MED23
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse, Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR