Anti-CECB1 Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
ROCK100-401-242EA 379 EUR
ROCK100-401-242
Anti-CECB1 Rabbit Polyclonal Antibody
Antikörper
Anti-cecropin B is useful for the detection of cecropin B in various immunological assays.  This reagent has specific value for the study of cecropins in transgenic plants. The antibody may also recognize naturally occurring cecropin A.

Recommended Dilutions: ELISA: 1:5000 - 1:20000; Western Blot: 1:500 - 1:5000; Immunoprecipitation: 1:100;

Type: Primary
Antigen: CECB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Silkworm
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR