Anti-DIABLO Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
ROCK100-401-244EA 379 EUR
ROCK100-401-244
Anti-DIABLO Rabbit Polyclonal Antibody
Antikörper
This antibody reacts with human Smac/DIABLO protein by immunoblot.  Lysates from human HeLa and LNCaP cells are positive for Smac/DIABLO.  Other animal tissues have not been tested.

Recommended Dilutions: ELISA: 1:5000 - 1:20000; Western Blot: 1:1000 - 1:2000; Immunoprecipitation: 1:100; Other Dilution: User Optimized

Type: Primary
Antigen: DIABLO
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR