Anti-BIVM Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
ROCK100-401-250EA 379 EUR
ROCK100-401-250
Anti-BIVM Rabbit Polyclonal Antibody
Antikörper
This polyclonal antibody reacts human BIVM at the C-terminal region and is useful for detection of BIVM by ELISA or western blotting.

Recommended Dilutions: ELISA: 1:5000 - 1:25000; Western Blot: 1:1000;

Type: Primary
Antigen: BIVM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR