Anti-RELA Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
ROCK100-401-264EA 379 EUR
ROCK100-401-264
Anti-RELA Rabbit Polyclonal Antibody
Antikörper
Phospho NFkB antibody reacts human pS276 p65 and shows minimal reactivity by western blot with non-phosphorylated p65 and minimal reactivity by ELISA against the non-phosphorylated form of the immunizing peptide.

Recommended Dilutions: ELISA: 1:10000 - 1:30,000; Western Blot: 1:1000 - 1:5000; Immunochemistry: 1:200-1:800;

Type: Primary
Antigen: RELA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR