Anti-NEU1 Rabbit Polyclonal Antibody

ROCK100-401-396EA 518 EUR
ROCK100-401-396
Anti-NEU1 Rabbit Polyclonal Antibody
This antibody is suitable for western blotting.

Recommended Dilutions: ELISA: 1:5000 - 1:25000; Immunofluorescence: 1:500; Western Blot: 1:500 - 1:2000;

Recommended Dilutions: ELISA: 1:5000 - 1:25000; Immunofluorescence: 1:500; Western Blot: 1:500 - 1:2000;

Type: Primary
Antigen: NEU1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR