Anti-AKT1 Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
ROCK100-401-401EA 402 EUR
ROCK100-401-401
Anti-AKT1 Rabbit Polyclonal Antibody
Antikörper
Primary Rabbit Anti-AKT Reacts with Chicken, Human, Mouse, Rat

Recommended Dilutions: ELISA: 1:2000 - 1:10000; Western Blot: 1:500 - 1:2000; Immunochemistry: 1:500 - 1:2000;

Type: Primary
Antigen: AKT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Chicken
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR