Anti-ANGPT2 Rabbit Polyclonal Antibody
Lieferant: Rockland Immunochemicals
ROCK100-401-402EA 783 EUR
ROCK100-401-402
Anti-ANGPT2 Rabbit Polyclonal Antibody
Antikörper
Angiopoietin 2 Antibody is suitable for western blotting and other antibody based assays.  A 1:500 dilution is recommended for western blotting.
Recommended Dilutions: ELISA: 1:5000 - 1:25000; Western Blot: 1:500 - 1:2000; Immunochemistry: 1:200-1:500;
Type: Primary
Antigen: ANGPT2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype:
Reactivity: Human, Mouse