Anti-NOTCH2 Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
ROCK100-401-406EA 402 EUR
ROCK100-401-406
Anti-NOTCH2 Rabbit Polyclonal Antibody
Antikörper
This antibody has been tested for use in ELISA and western blot.  A 1:500 dilution was effective for staining FFPE human kidney tissue by immunochemistry . Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: ELISA: 1:30,000 - 1:90,000; Western Blot: 1:400 - 1:2000; Immunochemistry: 1:200-1:800;

Type: Primary
Antigen: NOTCH2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype:
Reactivity: Human, Dog, Chimpanzee
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR