Anti-STAT4 Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
ROCK100-401-861EA 379 EUR
ROCK100-401-861
Anti-STAT4 Rabbit Polyclonal Antibody
Antikörper
This antibody has been tested for use in ELISA, western blotting and immunoprecipitation.  Reactivity in other immunoassays is unknown.  A HeLa whole cell lysate or mouse testis lysate are suitable for use as a positive control.

Recommended Dilutions: ELISA: 1:5000 - 1:25000; Western Blot: 1:5000; Immunoprecipitation: Use 5 µl;

Type: Primary
Antigen: STAT4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR