Anti-CUL9 Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
ROCK100-401-A07EA 379 EUR
ROCK100-401-A07
Anti-CUL9 Rabbit Polyclonal Antibody
Antikörper
This antibody reacts with human and mouse PARC by western blot and immunoprecipitation.  The antibody immunoprecipitates Protein from cell lysates (using HeLa, NIH-3T3, and others).  To date co-immuno-precipitation using 35S-IP has been negative.

Recommended Dilutions: ELISA: 1:2000 - 1:10000; Western Blot: 1:500 - 1:1000;

Type: Primary
Antigen: CUL9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR