Anti-DDB1 Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
ROCK100-401-A09EA 783 EUR
ROCK100-401-A09
Anti-DDB1 Rabbit Polyclonal Antibody
Antikörper
Anti-DDB1 antibody reacts with human and mouse  DDB1 by western blot and immunoprecipitation.  The antibody immunoprecipitates in vitro translated Protein and Protein from cell lysates (using HeLa, NIH-3T3, and others).

Recommended Dilutions: ELISA: 1:2000 - 1:10000; Western Blot: 1:500 - 1:1000; Immunoprecipitation: 1:500;

Type: Primary
Antigen: DDB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR