Anti-DDB2 Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
ROCK100-401-A10EA 783 EUR
ROCK100-401-A10
Anti-DDB2 Rabbit Polyclonal Antibody
Antikörper
This antibody reacts with human DDB2 by western blot and immunoprecipitation.  The antibody immunoprecipitates in vitro translated Protein and Protein from cell lysates (using 293T, and others).  Coimmunoprecipitation of related Proteins has not been tested.

Recommended Dilutions: ELISA: 1:2000 - 1:10000; Western Blot: 1:500 - 1:1000;

Type: Primary
Antigen: DDB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR