Anti-CAND1 Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
ROCK100-401-A11EA 379 EUR
ROCK100-401-A11
Anti-CAND1 Rabbit Polyclonal Antibody
Antikörper
This antibody reacts with human, rat  and mouse CAND1 by western blot and immunoprecipita-tion.  The antibody immunoprecipitates in vitro translated Protein and Protein from cell lysates (using HeLa and NIH-3T3, and others).

Recommended Dilutions: ELISA: 1:2000 - 1:10000; Western Blot: 1:500 - 1:1000;

Type: Primary
Antigen: CAND1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR