Anti-RNF7 Rabbit Monoclonal Antibody (Unconjugated) [clone: 14A11]

Lieferant: Rockland Immunochemicals
ROCK100-401-A14EA 379 EUR
ROCK100-401-A14
Anti-RNF7 Rabbit Monoclonal Antibody (Unconjugated) [clone: 14A11]
Antikörper
This antibody reacts with human ROC2 by western blot and immunoprecipitation.  The antibody immunoprecipitates in vitro translated Protein and Protein from overexpressing cell lysates (using HeLa and NIH-3T3, and others).

Recommended Dilutions: ELISA: 1:2000 - 1:10000; Western Blot: 1:500 - 1:1000;

Type: Primary
Antigen: RNF7
Clonality: Monoclonal
Clone: 14A11
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR