Anti-ANAPC11 Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
ROCK100-401-A15EA 379 EUR
ROCK100-401-A15
Anti-ANAPC11 Rabbit Polyclonal Antibody
Antikörper
This antibody reacts with human APC11 by western blot and immunoprecipitation.  The antibody immunoprecipitates in vitro translated Protein and Protein from overexpressing cell lysates (using HeLa and NIH-3T3, and others).

Recommended Dilutions: ELISA: 1:2000 - 1:10000; Western Blot: 1:500 - 1:1000;

Type: Primary
Antigen: ANAPC11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR