Anti-CDC20 Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
ROCK100-401-A17EA 379 EUR
ROCK100-401-A17
Anti-CDC20 Rabbit Polyclonal Antibody
Antikörper
This antibody reacts with human CDC20 (fizzy) by western blot and immunoprecipitation.  The antibody immunoprecipitates in vitro translated Protein and Protein from overexpressing cell lysates (using HeLa and NIH-3T3, and others).

Recommended Dilutions: ELISA: 1:2000 - 1:10000; Western Blot: 1:500 - 1:1000;

Type: Primary
Antigen: CDC20
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR