Anti-PhoA Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
ROCK100-4134EA 1950 EUR
ROCK100-4134
Anti-PhoA Rabbit Polyclonal Antibody
Antikörper
Anti-Alkaline Phosphatase (E. Coli) Antibody has been assayed against 1.0 µg of Alkaline Phosphatase (E.coli) in a standard ELISA using Peroxidase conjugated Affinity Purified anti-Rabbit IgG (H&L) (Goat) and ABTS (2,2’-azino-bis-(3-ethylbenthiazoline-6-sulfonic acid))

Recommended Dilutions: ELISA: 1:8000 to 1:40000; Western Blot: 1:500 - 1:2000;
Restore with deionized water (or equivalent); 2.0 ml

Type: Primary
Antigen: PhoA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: E. coli
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR