Anti-GLB1 Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
ROCK100-4136EA 239 EUR
ROCK100-4136
Anti-GLB1 Rabbit Polyclonal Antibody
Antikörper
Primary Rabbit Anti-BETA GALACTOSIDASE (E.coli) Reacts with Pan

Type: Primary
Antigen: GLB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: E. coli
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR