Anti-AMY Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
ROCK100-4145EA 1950 EUR
ROCK100-4145
Anti-AMY Rabbit Polyclonal Antibody
Antikörper
Primary Rabbit Anti-ALPHA AMYLASE (Bacillus amyloliquefaciens) Reacts with Pan

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunoprecipitation: 1:100;
Restore with deionized water (or equivalent); 2.0 ml

Type: Primary
Antigen: AMY
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bacillus amyloliquefaciens
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR