Anti-CPA1 Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
ROCK100-4152EA 1020 EUR
ROCK100-4152
Anti-CPA1 Rabbit Polyclonal Antibody
Antikörper
Carboxypeptidase A Antibody has been tested by ELISA and western blot. Optimization for specific immunoassays should be determined by the researcher.

Recommended Dilutions: ELISA: 1:5000 - 1:25000; Western Blot: 1:500 - 1:3000;
Restore with deionized water (or equivalent); 2.0 ml

Type: Primary
Antigen: CPA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Yeast
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR