Anti-DLD Rabbit Monoclonal Antibody (Unconjugated) [clone: 7C5G1H6H10]

Lieferant: Rockland Immunochemicals
ROCK100-4160EA 1950 EUR
ROCK100-4160
Anti-DLD Rabbit Monoclonal Antibody (Unconjugated) [clone: 7C5G1H6H10]
Antikörper
This product has been assayed against 1.0 µg of Lipoamide Dehydrogenase (Porcine Heart) in a standard ELISA using Peroxidase conjugated Affinity Purified anti-Rabbit IgG (H&L) (Goat) and ABTS (2,2’-azino-bis-(3-ethylbenthiazoline-6-sulfonic acid))

Recommended Dilutions: ELISA: 1:5000 - 1:20000; Western Blot: 1:500 - 1:5000; Immunoprecipitation: 1:100;
Restore with deionized water (or equivalent); 2.0 ml

Type: Primary
Antigen: DLD
Clonality: Monoclonal
Clone: 7C5G1H6H10
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Pig
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR