Anti-PEPC Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
ROCK100-4163EA 1020 EUR
ROCK100-4163
Anti-PEPC Rabbit Polyclonal Antibody
Antikörper
Primary Rabbit Anti-PHOSPHO ENOL PYRUVATE CARBOXYLASE (Maize Leaves) Reacts with Pan

Recommended Dilutions: ELISA: 1:30,000 - 1:160,000; Western Blot: 1:5000 - 1:20000;
Restore with deionized water (or equivalent); 2.0 ml

Type: Primary
Antigen: PEPC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Maize Leaves, Maize
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR