Anti-RELA Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
ROCK100-4165EA 783 EUR
ROCK100-4165
Anti-RELA Rabbit Polyclonal Antibody
Antikörper
Anti-NFkB p65 (Rel A) is suitable for the detection by immunoblot of human and mouse NFkB p65 (Rel A), immunochemistry, and ELISA.  This product was also tested in a gel supershift assay and found to be reactive against all p65 (Rel A).

Recommended Dilutions: ELISA: 1:5000; GelShift: 05 µl - 1.0 µl; Western Blot: 1:2000 - 1:5000; Immunochemistry: 1:400;

Type: Primary
Antigen: RELA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype:
Reactivity: Mouse, Rat, Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR