Anti-RELA Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
ROCK100-4165NEA 379 EUR
ROCK100-4165N
Anti-RELA Rabbit Polyclonal Antibody
Antikörper
Anti-NFkB p65 (N-terminal specific) is suitable for the detection by immunoblot of human NFkB p65 (N-terminal specific).  Functionality in supershift assays has not been determined.

Recommended Dilutions: ELISA: 1:5000 - 1:25000; Western Blot: 1:5000 - 1:10000;

Type: Primary
Antigen: RELA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR