Anti-NFKBIA Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
ROCK100-4167CEA 379 EUR
ROCK100-4167C
Anti-NFKBIA Rabbit Polyclonal Antibody
Antikörper
Anti-IkBa Antibody is suitable for western blotting, ELISA, and IHC. Researchers should determine optimal titers for applications that are not stated below.

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;

Type: Primary
Antigen: NFKBIA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype:
Reactivity: Mouse, Rat, Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR