Anti-CELA1 Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
ROCK100-4168EA 1020 EUR
ROCK100-4168
Anti-CELA1 Rabbit Polyclonal Antibody
Antikörper
Elastase antibody has been teted for functionality in ELISA and western blot. Usage in other immunoassays such as immunohistochemisty is expected but has not been determined. Optimal concentrations should be determined by researcher.

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000;
Restore with deionized water (or equivalent); 2.0 ml

Type: Primary
Antigen: CELA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Pig
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR