Anti-LYZ Rabbit Monoclonal Antibody (Unconjugated) [clone: 17D10]

Lieferant: Rockland Immunochemicals
ROCK100-4172EA 1020 EUR
ROCK100-4172
Anti-LYZ Rabbit Monoclonal Antibody (Unconjugated) [clone: 17D10]
Antikörper
Anti-LYSOZYME antibody has been assayed against 1.0 µg of Lysozyme (Hen Egg White) in a standard ELISA. A working dilution of 1:1000 to 1:2000 of the reconstitution concentration is suggested for this product.

Recommended Dilutions: ELISA: 1:5000 - 1:20000; Western Blot: 1:500 - 1:5000; Immunoprecipitation: 1:100;
Restore with deionized water (or equivalent); 2.0 ml

Type: Primary
Antigen: LYZ
Clonality: Monoclonal
Clone: 17D10
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Chicken
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR