Anti-RNASE1 Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
ROCK100-4188EA 1020 EUR
ROCK100-4188
Anti-RNASE1 Rabbit Polyclonal Antibody
Antikörper
Anti-Ribonuclease A Antibody is suitable for western blotting, IP and for ELISA. Researchers should determine optimal titers for applications that are not stated below.

Recommended Dilutions: ELISA: 1:30,000 - 1:150000; Western Blot: 1:3000 - 1:15000; Immunoprecipitation: 1:100;
Restore with deionized water (or equivalent); 2.0 ml

Type: Primary
Antigen: RNASE1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR