Anti-SOD1 Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
ROCK100-4191EA 1020 EUR
ROCK100-4191
Anti-SOD1 Rabbit Polyclonal Antibody
Antikörper
Primary Rabbit Anti-SUPEROXIDE DISMUTASE (Bovine Erythrocytes) Reacts with Pan

Recommended Dilutions: ELISA: 1:6000 - 1:30,000; Western Blot: 1:600 - 1:3000;
Restore with deionized water (or equivalent); 2.0 ml

Type: Primary
Antigen: SOD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR