Anti-B-Phycoerythrin Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
ROCK100-4199EA 708 EUR
ROCK100-4199
Anti-B-Phycoerythrin Rabbit Polyclonal Antibody
Antikörper
Suitable for immunoblotting (western or dot blot), ELISA, immunoprecipitation and most immunological methods requiring high titer and specificity.

Recommended Dilutions: ELISA: 1:20000-1:100000; Western Blot: 1:2000-1:10000; Immunochemistry: 1:1000-1:5000
Restore with deionized water (or equivalent); 2.0 ml

Type: Tag
Antigen: B-Phycoerythrin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: pan
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR