Anti-GPT Sheep Polyclonal Antibody

Lieferant: Rockland Immunochemicals
ROCK100-601-206EA 1020 EUR
ROCK100-601-206
Anti-GPT Sheep Polyclonal Antibody
Antikörper
This product has been assayed against 1.0 µg of Alanine Transaminase (ALT) (Pig Heart) in a standard ELISA using Peroxidase conjugated Affinity Purified anti-Sheep IgG (H&L) (Goat) and ABTS (2,2’-azino-bis-(3-ethylbenthiazoline-6-sulfonic acid))

Recommended Dilutions: ELISA: 1:5000 - 1:25000; Western Blot: 1:500 - 1:3000;
Restore with deionized water (or equivalent); 2.0 ml

Type: Primary
Antigen: GPT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype:
Reactivity: Pig
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR