Anti-faoxC Sheep Polyclonal Antibody
Lieferant: Rockland Immunochemicals
ROCK100-601-236EA 1950 EUR
ROCK100-601-236
Anti-faoxC Sheep Polyclonal Antibody
Antikörper
This product has been assayed against 1.0 µg of Fructosyl-Amino Acid Oxidase (E.coli) in a standard ELISA using Peroxidase conjugated Affinity Purified anti-Sheep IgG (H&L) (Rabbit) and ABTS (2,2’-azino-bis-(3-ethylbenthiazoline-6-sulfonic acid))
Recommended Dilutions: ELISA: 1:5000 - 1:20000; Western Blot: 1:500 - 1:5000; Immunoprecipitation: 1:100;
Restore with deionized water (or equivalent); 2.0 ml
Type: Primary
Antigen: faoxC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype:
Reactivity: Corynebacterium Sp, Corynebacterium