Anti-BSA Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
ROCK101-4133EA 482 EUR
ROCK101-4133
Anti-BSA Rabbit Polyclonal Antibody
Antikörper
Anti-ALB antibody is suitable for western blotting, IHC and ELISA. The antibody may be suitable for other applications, although no supporting data is available.

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;
Restore with deionized water (or equivalent); 2.0 ml

Type: Primary
Antigen: BSA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR