Anti-IgM µ Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
ROCK102-4107EA 128 EUR
ROCK102-4107
Anti-IgM µ Rabbit Polyclonal Antibody
Antikörper
Secondary Rabbit Anti-IgM mu chain Reacts with Cat (Feline)

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;
Restore with deionized water (or equivalent); 2.0 ml

Type: Secondary
Antigen: IgM µ
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: µ-chain
Host: Rabbit
Isotype: IgM
Reactivity: Cat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR