Anti-Goat serum Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
ROCK105-4101EA 249 EUR
ROCK105-4101
Anti-Goat serum Rabbit Polyclonal Antibody
Antikörper
Anti-goat serum antibody is suitable for western blotting, IP and for ELISA. Researchers should determine optimal titers for applications that are not stated below.

Recommended Dilutions:
Restore with deionized water (or equivalent); 2.0 ml

Type: Primary
Antigen: Goat serum
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Goat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR