Anti-Guinea Pig serum Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
ROCK106-4101EA 493 EUR
ROCK106-4101
Anti-Guinea Pig serum Rabbit Polyclonal Antibody
Antikörper
Primary Rabbit Anti-GUINEA PIG SERUM Reacts with Guinea Pig

Recommended Dilutions:
Restore with deionized water (or equivalent); 2.0 ml

Type: Primary
Antigen: Guinea Pig serum
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Guinea Pig
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR