Anti-Golden Syrian Hamster serum Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
ROCK107-4101EA 482 EUR
ROCK107-4101
Anti-Golden Syrian Hamster serum Rabbit Polyclonal Antibody
Antikörper
Primary Rabbit Anti-GOLDEN SYRIAN HAMSTER SERUM Reacts with Hamster

Recommended Dilutions:
Restore with deionized water (or equivalent); 2.0 ml

Type: Primary
Antigen: Golden Syrian Hamster serum
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Hamster, Golden Syrian Hamster
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR