Anti-CRP Goat Polyclonal Antibody

Lieferant: Rockland Immunochemicals
ROCK109-1132EA 482 EUR
ROCK109-1132
Anti-CRP Goat Polyclonal Antibody
Antikörper
Primary Goat Anti-HUMAN CRP Reacts with Human

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;
Restore with deionized water (or equivalent); 2.0 ml

Type: Primary
Antigen: CRP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR