Anti-IL1B Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
ROCK109-401-301EA 1020 EUR
ROCK109-401-301
Anti-IL1B Rabbit Polyclonal Antibody
Antikörper
Anti-Human IL-1-beta has been tested for use in neutralizations, ELISA, radioimmunoassays, flow cytometry, immunochemistry, immunoblotting and immunoprecipitation. It recognizes the 17,000 MW mature IL-1-beta.

Recommended Dilutions: ELISA: 1:500 - 1:2000; Neutralization: 1:100; Western Blot: 1:1000; Immunochemistry: 1:100 - 1:200; Immunoprecipitation: 1:400 - 1:800;

Type: Primary
Antigen: IL1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Human, Primate, Dog
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR