Anti-IL1R1 Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
ROCK109-401-304EA 1020 EUR
ROCK109-401-304
Anti-IL1R1 Rabbit Polyclonal Antibody
Antikörper
This antiserum against anti-Human IL-1 Receptor Type I (IL-1RI) has been tested for use in ELISA,  radioimmunoassays and immunoprecipitation.  Reactivity in other immunoassays is unknown.

Recommended Dilutions:

Type: Primary
Antigen: IL1R1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Primate
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR