Anti-IL1R2 Rabbit Monoclonal Antibody (Unconjugated) [clone: 14H5]

Lieferant: Rockland Immunochemicals
ROCK109-401-305EA 1020 EUR
ROCK109-401-305
Anti-IL1R2 Rabbit Monoclonal Antibody (Unconjugated) [clone: 14H5]
Antikörper
This antiserum against Human IL-1 Receptor Type II (IL-1RII) has been tested for use in ELISA, radioimmunoassays and immunoprecipitation.  Reactivity in other immunoassays is unknown.  IL-1RII has been termed p68 or CD121 in some laboratories.

Recommended Dilutions: ELISA: 1:1000 - 1:2000; Western Blot: 1:500 - 1:2000; Immunoprecipitation: 1:400 - 1:800;

Type: Primary
Antigen: IL1R2
Clonality: Monoclonal
Clone: 14H5
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Primate
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR